ENAH38A2D9D8 Enamine

benzyl 1-oxo-7-azaspiro[3.5]nonane-7-carboxylate

  • MDL number MFCD31915146
  • Cas Number : 166306-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352353
Availability details for
Quantity
  • Availability
  • Add to Cart