ENAH38A2D9DA Enamine

5-(4-methoxyphenyl)-2-methyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD16326225
  • Cas Number : 1007874-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352368
Availability details for
Quantity
  • Availability
  • Add to Cart