ENAH38A2D9DB Enamine

2-{[4-(trifluoromethoxy)phenyl]methyl}pyrrolidine hydrochloride

  • MDL number MFCD31915200
  • Cas Number : 2309455-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352370
Availability details for
Quantity
  • Availability
  • Add to Cart