ENAH38A2D9DC Enamine

ethyl 2-(aminomethyl)-6,6-difluorospiro[3.3]heptane-2-carboxylate hydrochloride

  • MDL number MFCD31915246
  • Cas Number : 2309447-18-3
  • Molecular Weight : 269.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352380
Availability details for
Quantity
  • Availability
  • Add to Cart