ENAH38A2D9DE Enamine

3-propyl-1H-indazole-5-carboxylic acid

  • MDL number MFCD24499971
  • Cas Number : 1197943-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352388
Availability details for
Quantity
  • Availability
  • Add to Cart