ENAH38A2D9E3 Enamine

[1-(3,6-dihydro-2H-pyran-4-yl)cyclobutyl]methanamine hydrochloride

  • MDL number MFCD31915248
  • Cas Number : 2309474-37-9
  • Molecular Weight : 203.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352405
Availability details for
Quantity
  • Availability
  • Add to Cart