ENAH38A2D9E6 Enamine

N-(azetidin-3-yl)ethane-1-sulfonamide; trifluoroacetic acid

  • MDL number MFCD31915204
  • Cas Number : 2309456-99-1
  • Molecular Weight : 278.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352444
Availability details for
Quantity
  • Availability
  • Add to Cart