ENAH38A2D9E7 Enamine

[(azetidin-3-yl)sulfamoyl]dimethylamine; trifluoroacetic acid

  • MDL number MFCD31915205
  • Cas Number : 2309474-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352445
Availability details for
Quantity
  • Availability
  • Add to Cart