ENAH38A2D9E8 Enamine

2-(ethanesulfonyl)pyrimidin-5-amine hydrochloride

  • MDL number MFCD31915249
  • Cas Number : 2309466-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352447
Availability details for
Quantity
  • Availability
  • Add to Cart