ENAH38A2D9EC Enamine

methyl (3R)-3-amino-3-(4-phenylphenyl)propanoate

  • MDL number MFCD08056834
  • Cas Number : 327617-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352478
Availability details for
Quantity
  • Availability
  • Add to Cart