ENAH38A2D9EE Enamine

tert-butyl 6-[(chlorosulfonyl)methyl]-5-oxa-2-azaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD31915208
  • Cas Number : 2309466-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352497
Availability details for
Quantity
  • Availability
  • Add to Cart