ENAH38A2D9F3 Enamine

(2S,4R)-2-(chloromethyl)-4-methoxypyrrolidine hydrochloride

  • MDL number MFCD31915210
  • Cas Number : 2309433-08-5
  • Molecular Weight : 186.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352555
Availability details for
Quantity
  • Availability
  • Add to Cart