ENAH38A2D9F8 Enamine

tert-butyl 6-(aminomethyl)-5-oxa-8-azaspiro[3.5]nonane-8-carboxylate

  • MDL number MFCD31915212
  • Cas Number : 2169022-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352580
Availability details for
Quantity
  • Availability
  • Add to Cart