ENAH38A2D9FD Enamine

4-[(3-methylphenyl)amino]pyrimidine-5-carboxylic acid

  • MDL number MFCD27945623
  • Cas Number : 16100-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352593
Availability details for
Quantity
  • Availability
  • Add to Cart