ENAH38A2DA07 Enamine

4-(1,1-difluoroethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD28534978
  • Cas Number : 1782576-12-8
  • Molecular Weight : 240.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352642
Availability details for
Quantity
  • Availability
  • Add to Cart