ENAH38A2DA0B Enamine

ethyl 4-(2,3-dihydro-1-benzofuran-5-yl)-2,4-dioxobutanoate

  • MDL number MFCD31915270
  • Cas Number : 2309463-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352650
Availability details for
Quantity
  • Availability
  • Add to Cart