ENAH38A2DA0F Enamine

N-[5-(4-bromophenyl)-1,3,4-oxadiazol-2-yl]-2,2-dimethylpropanamide

  • MDL number MFCD07732243
  • Cas Number : 905680-01-5
  • Molecular Weight : 324.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352678

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352678
Availability details for
Quantity
  • Availability
  • Add to Cart