ENAH38A2DA15 Enamine

2-amino-4H,5H,6H,7H,8H-cyclohepta[d][1,3]oxazol-4-one

  • MDL number MFCD31915272
  • Cas Number : 2309475-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352714
Availability details for
Quantity
  • Availability
  • Add to Cart