ENAH38A2DA2D Enamine

1-(bromomethyl)-1-methanesulfonylcyclopropane

  • MDL number MFCD31915333
  • Cas Number : 2091872-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352842
Availability details for
Quantity
  • Availability
  • Add to Cart