ENAH38A2DA32 Enamine

1-[2-chloro-4-(4-methyl-1,3-thiazol-5-yl)phenyl]ethan-1-one

  • MDL number MFCD31915286
  • Cas Number : 2019353-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352889
Availability details for
Quantity
  • Availability
  • Add to Cart