ENAH38A2DA37 Enamine

4-[(2-methoxyphenyl)amino]pyrimidine-5-carboxylic acid

  • MDL number MFCD11573896
  • Cas Number : 1082361-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352921
Availability details for
Quantity
  • Availability
  • Add to Cart