ENAH38A2DA38 Enamine

4-[(3-cyanophenyl)amino]pyrimidine-5-carboxylic acid

  • MDL number MFCD26652799
  • Cas Number : 1507757-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352922
Availability details for
Quantity
  • Availability
  • Add to Cart