ENAH38A2DA40 Enamine

(5-methyl-4,5-dihydro-1,2-oxazol-3-yl)methanesulfonyl chloride

  • MDL number MFCD31915378

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353015
Availability details for
Quantity
  • Availability
  • Add to Cart