ENAH38A2DA4A Enamine

5-(azetidin-3-yl)-1-methyl-1H-pyrazole-4-carboxylic acid dihydrochloride

  • MDL number MFCD31915295
  • Cas Number : 2309447-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353086
Availability details for
Quantity
  • Availability
  • Add to Cart