ENAH38A2DA4C Enamine

tert-butyl 5-(chloromethyl)-2,3-dihydro-1H-isoindole-2-carboxylate

  • MDL number MFCD31915386
  • Cas Number : 2309462-02-8
  • Molecular Weight : 267.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353117

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353117
Availability details for
Quantity
  • Availability
  • Add to Cart