ENAH38A2DA4E Enamine

3-aminooxolane-3-carboxylic acid hydrochloride

  • MDL number MFCD11035895
  • Cas Number : 919098-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353169
Availability details for
Quantity
  • Availability
  • Add to Cart