ENAH38A2DA51 Enamine

2-(2-chloroethoxy)-2-methylpropanenitrile

  • MDL number MFCD19232778
  • Cas Number : 870562-14-4
  • Molecular Weight : 147.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353186
Availability details for
Quantity
  • Availability
  • Add to Cart