ENAH38A2DA56 Enamine

3-chloro-5H,6H,7H,8H-imidazo[1,2-b]pyridazine

  • MDL number MFCD31915296
  • Cas Number : 2309474-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353246
Availability details for
Quantity
  • Availability
  • Add to Cart