ENAH38A2DA5A Enamine

5-(chloromethyl)-1-ethyl-1H-1,2,3-benzotriazole

  • MDL number MFCD31915297
  • Cas Number : 2309461-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353328
Availability details for
Quantity
  • Availability
  • Add to Cart