ENAH38A2DA5D Enamine

8,8-difluoro-4-azabicyclo[5.1.0]octan-3-one

  • MDL number MFCD31915394
  • Cas Number : 2309466-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353342
Availability details for
Quantity
  • Availability
  • Add to Cart