ENAH38A2DA5E Enamine

sodium 4-cyanopyridine-2-sulfinate

  • MDL number MFCD31915298
  • Cas Number : 2274738-39-3
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353343
Availability details for
Quantity
  • Availability
  • Add to Cart