ENAH38A2DA67 Enamine

4-methoxy-1,3-oxazolidin-2-one

  • MDL number MFCD17170082
  • Cas Number : 14441-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353458
Availability details for
Quantity
  • Availability
  • Add to Cart