ENAH38A2DA6D Enamine

6-chloro-2-[(6-methylpyridin-3-yl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD25627915
  • Cas Number : 1535166-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353510
Availability details for
Quantity
  • Availability
  • Add to Cart