ENAH38A2DA6F Enamine

ethyl 2-chloro-6-methoxypyridine-4-carboxylate

  • MDL number MFCD14698195
  • Cas Number : 106719-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353522
Availability details for
Quantity
  • Availability
  • Add to Cart