ENAH38A2DA70 Enamine

1-benzyl-3-iodobenzene

  • MDL number MFCD31915452
  • Cas Number : 92903-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353527
Availability details for
Quantity
  • Availability
  • Add to Cart