ENAH38A2DA77 Enamine

benzyl 3-(2-aminoethyl)-3-methoxyazetidine-1-carboxylate hydrochloride

  • MDL number MFCD31915458
  • Cas Number : 2309462-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353611
Availability details for
Quantity
  • Availability
  • Add to Cart