ENAH38A2DA83 Enamine

5-(hydroxymethyl)azepan-4-ol; trifluoroacetic acid

  • MDL number MFCD31915312
  • Cas Number : 2309468-22-0
  • Molecular Weight : 259.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353672
Availability details for
Quantity
  • Availability
  • Add to Cart