ENAH38A2DA89 Enamine

5-fluoro-1H-1,3-benzodiazol-6-amine dihydrochloride

  • MDL number MFCD31915501
  • Cas Number : 2309454-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353693
Availability details for
Quantity
  • Availability
  • Add to Cart