ENAH38A2DA8B Enamine

5-(tert-butoxy)pyridine-3-carboxylic acid

  • MDL number MFCD31915502
  • Cas Number : 2309467-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353725
Availability details for
Quantity
  • Availability
  • Add to Cart