ENAH38A2DA90 Enamine

4-[4-(trifluoromethyl)-1H-1,2,3-triazol-1-yl]piperidine

  • MDL number MFCD31915540
  • Cas Number : 2309459-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353816
Availability details for
Quantity
  • Availability
  • Add to Cart