ENAH38A2DA92 Enamine

2-phenoxycyclohexan-1-one

  • MDL number MFCD08836171
  • Cas Number : 14004-19-4
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353831
Availability details for
Quantity
  • Availability
  • Add to Cart