ENAH38A2DA98 Enamine

4-[(4-cyclopropylpiperazin-1-yl)methyl]aniline trihydrochloride

  • MDL number MFCD32066404
  • Cas Number : 2309453-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353844
Availability details for
Quantity
  • Availability
  • Add to Cart