ENAH38A2DA99 Enamine

{4-methyl-2-oxabicyclo[2.2.1]heptan-1-yl}methanol

  • MDL number MFCD31915319
  • Cas Number : 2309461-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353849
Availability details for
Quantity
  • Availability
  • Add to Cart