ENAH38A2DAA8 Enamine

6-chloro-2-[(3-methylphenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD18586588
  • Cas Number : 1374969-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353960
Availability details for
Quantity
  • Availability
  • Add to Cart