ENAH38A2DAA9 Enamine

6-chloro-2-[(3-fluorophenyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD18570380
  • Cas Number : 1406100-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7353961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7353961
Availability details for
Quantity
  • Availability
  • Add to Cart