ENAH38A2DACC Enamine

tert-butyl (3R)-3-amino-3-(2,6-difluorophenyl)propanoate hydrochloride

  • MDL number MFCD31915330
  • Cas Number : 2309433-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354166
Availability details for
Quantity
  • Availability
  • Add to Cart