ENAH38A2DAD0 Enamine

5-azaspiro[3.5]nonane-8-carboxylic acid hydrochloride

  • MDL number MFCD31915331
  • Cas Number : 2309448-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354185
Availability details for
Quantity
  • Availability
  • Add to Cart