ENAH38A2DAE5 Enamine

8-oxa-3-azaspiro[5.6]dodecane hydrochloride

  • MDL number MFCD31915735
  • Cas Number : 2309469-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354272
Availability details for
Quantity
  • Availability
  • Add to Cart