ENAH38A2DAEF Enamine

(2E)-3-(4-methoxyphenyl)pent-2-enoic acid

  • MDL number MFCD09889443
  • Cas Number : 30219-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354338
Availability details for
Quantity
  • Availability
  • Add to Cart