ENAH38A2DAF9 Enamine

(2S,3S)-2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)-3-methylpentanoic acid

  • MDL number MFCD31915798
  • Cas Number : 53481-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7354368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354368
Availability details for
Quantity
  • Availability
  • Add to Cart